ENA024294288 Enamine

1-{[3,5-bis(methoxycarbonyl)-4-methylthien-2-yl]sulfonyl}piperidine-4-carboxylic acid

  • MDL number MFCD04625505
  • Cas Number : 731003-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08248
Availability details for
Quantity
  • Availability
  • Add to Cart