ENA024294294 Enamine

4-[4-(benzenesulfonyl)piperazin-1-yl]-4-oxobutanoic acid

  • MDL number MFCD03989648
  • Cas Number : 565181-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-110
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08254
Availability details for
Quantity
  • Availability
  • Add to Cart