ENA024294303 Enamine

2-chloro-1-{1-[2-(4-chlorophenyl)ethyl]-2,5-dimethyl-1H-pyrrol-3-yl}ethanone

  • MDL number MFCD04606969
  • Cas Number : 565198-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08263

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08263
Availability details for
Quantity
  • Availability
  • Add to Cart