ENA024294304 Enamine

2-chloro-1-{1-[2-(4-fluorophenyl)ethyl]-2,5-dimethyl-1H-pyrrol-3-yl}ethan-1-one

  • MDL number MFCD04606908
  • Cas Number : 556006-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08264
Availability details for
Quantity
  • Availability
  • Add to Cart