ENA024294311 Enamine

1-(thien-2-ylsulfonyl)piperazine

  • MDL number MFCD03987297
  • Cas Number : 565178-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08271

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08271
Availability details for
Quantity
  • Availability
  • Add to Cart