ENA024294315 Enamine

1-{[4-chloro-3-(trifluoromethyl)phenyl]sulfonyl}piperazine

  • MDL number MFCD04629611
  • Cas Number : 97630-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292...295
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08275
Availability details for
Quantity
  • Availability
  • Add to Cart