ENA024294319 Enamine

5-(2-hydroxybenzoyl)-1-(2-methoxyethyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD04621697
  • Cas Number : 730951-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08279
Availability details for
Quantity
  • Availability
  • Add to Cart