ENA024294324 Enamine

5-(4-chlorophenyl)-4-(3-ethoxypropyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03947450
  • Cas Number : 849587-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08284
Availability details for
Quantity
  • Availability
  • Add to Cart