ENA024294332 Enamine

5-(2-fluorophenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00298361
  • Cas Number : 311784-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08292

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08292
Availability details for
Quantity
  • Availability
  • Add to Cart