ENA024294361 Enamine

7-chloro-3-cyclopentyl-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD04627357
  • Cas Number : 731802-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08321
Availability details for
Quantity
  • Availability
  • Add to Cart