ENA024294374 Enamine

4-amino-5-(2,3-dihydro-1,4-benzodioxin-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02227482
  • Cas Number : 364599-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08334
Availability details for
Quantity
  • Availability
  • Add to Cart