ENA024294382 Enamine

4-(pyrrolidine-1-sulfonyl)benzohydrazide

  • MDL number MFCD04614717
  • Cas Number : 735322-66-4
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08342
Availability details for
Quantity
  • Availability
  • Add to Cart