ENA024294385 Enamine

4-(1H-indol-3-yl)-1,3-thiazol-2-amine

  • MDL number MFCD00449605
  • Cas Number : 22258-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08345
Availability details for
Quantity
  • Availability
  • Add to Cart