ENA024294386 Enamine

2-[2-methyl-5-(propan-2-yl)phenoxy]acetic acid

  • MDL number MFCD01365737
  • Cas Number : 19728-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08346
Availability details for
Quantity
  • Availability
  • Add to Cart