ENA024294387 Enamine

2-[4-(4-methylphenyl)-2-sulfanylidene-2,3-dihydro-1,3-thiazol-3-yl]acetic acid

  • MDL number MFCD03973221
  • Cas Number : 554407-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08347
Availability details for
Quantity
  • Availability
  • Add to Cart