ENA024294400 Enamine

2-[(2,4-dichlorobenzoyl)amino]-4-(methylthio)butanoic acid

  • MDL number MFCD04621497
  • Cas Number : 65054-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08360
Availability details for
Quantity
  • Availability
  • Add to Cart