ENA024294401 Enamine

4-(methylthio)-2-[(3,4,5-trimethoxybenzoyl)amino]butanoic acid

  • MDL number MFCD01250131
  • Cas Number : 55652-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08361

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08361
Availability details for
Quantity
  • Availability
  • Add to Cart