ENA024294408 Enamine

2-[(2-chloro-5-nitrobenzoyl)amino]-3-methylbutanoic acid

  • MDL number MFCD04619508
  • Cas Number : 1396968-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08368

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08368
Availability details for
Quantity
  • Availability
  • Add to Cart