ENA024294415 Enamine

2-[(2-chloro-4-nitrophenyl)formamido]-3-methylbutanoic acid

  • MDL number MFCD04619506
  • Cas Number : 1397003-81-4
  • Molecular Weight : 300.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08375
Availability details for
Quantity
  • Availability
  • Add to Cart