ENA024294426 Enamine

2-({[2-phenylvinyl]sulfonyl}amino)propanoic acid

  • MDL number MFCD03987303

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58...60
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08386
Availability details for
Quantity
  • Availability
  • Add to Cart