ENA024294430 Enamine

3-methyl-1-phenyl-5-pyrrolidin-1-yl-1H-pyrazole-4-carbaldehyde oxime

  • MDL number MFCD04621464
  • Cas Number : 882226-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08390

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08390
Availability details for
Quantity
  • Availability
  • Add to Cart