ENA024294444 Enamine

ethyl 2-amino-5-methyl-4-(4-methylphenyl)thiophene-3-carboxylate

  • MDL number MFCD01993633
  • Cas Number : 350997-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08404
Availability details for
Quantity
  • Availability
  • Add to Cart