ENA024294446 Enamine

({[4-chloro-3-(trifluoromethyl)phenyl]sulfonyl}amino)acetic acid

  • MDL number MFCD03619019
  • Cas Number : 612043-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08406
Availability details for
Quantity
  • Availability
  • Add to Cart