ENA024294449 Enamine

2-[2-(naphthalen-1-yl)acetamido]acetic acid

  • MDL number MFCD04627342
  • Cas Number : 6277-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08409
Availability details for
Quantity
  • Availability
  • Add to Cart