ENA024294455 Enamine

(2S)-2-{[(2E)-3-(4-methoxyphenyl)prop-2-enoyl]amino}propanoic acid

  • MDL number MFCD02323134
  • Cas Number : 1163681-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08415

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08415
Availability details for
Quantity
  • Availability
  • Add to Cart