ENA024294505 Enamine

2-chloro-1-(4-methoxyphenyl)propan-1-one

  • MDL number MFCD03428413
  • Cas Number : 81112-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08465
Availability details for
Quantity
  • Availability
  • Add to Cart