ENA024294525 Enamine

5-(4-fluorophenyl)thieno[2,3-d]pyrimidine-4-thiol

  • MDL number MFCD01917787
  • Cas Number : 356569-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08485
Availability details for
Quantity
  • Availability
  • Add to Cart