ENA024294526 Enamine

3-[3-(morpholin-4-yl)propyl]-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD04627359
  • Cas Number : 146381-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08486
Availability details for
Quantity
  • Availability
  • Add to Cart