ENA024294529 Enamine

2-(1,3-benzothiazol-2-yl)-4-chloro-3-methylbut-2-enenitrile

  • Cas Number : 735342-48-0
  • Molecular Weight : 248.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08489
Availability details for
Quantity
  • Availability
  • Add to Cart