ENA024294553 Enamine

2-(4-acetylphenoxy)-N-phenylacetamide

  • MDL number MFCD01733463
  • Cas Number : 17172-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08513
Availability details for
Quantity
  • Availability
  • Add to Cart