ENA024294555 Enamine

2-methanesulfonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD03021424
  • Cas Number : 1007984-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-133
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08515
Availability details for
Quantity
  • Availability
  • Add to Cart