ENA024294559 Enamine

4-methyl-2-morpholin-4-yl-1,3-thiazole-5-carbohydrazide

  • MDL number MFCD05263185
  • Cas Number : 743444-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08519

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08519
Availability details for
Quantity
  • Availability
  • Add to Cart