ENA024294560 Enamine

2-(morpholin-4-yl)-1,3-thiazole-4-carbohydrazide

  • MDL number MFCD05263198
  • Cas Number : 743444-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08520
Availability details for
Quantity
  • Availability
  • Add to Cart