ENA024294567 Enamine

1-(2,6-dichlorobenzyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD05263184
  • Cas Number : 956714-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08527
Availability details for
Quantity
  • Availability
  • Add to Cart