ENA024294572 Enamine

5-amino-3-[3-(methylamino)propyl]-1H-pyrazole-4-carbonitrile

  • MDL number MFCD05263126
  • Cas Number : 113513-30-7
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 127-128
Iupac Name 5-amino-3-[3-(methylamino)propyl]-1H-pyrazole-4-carbonitrile
Inchi Code 1S/C8H13N5/c1-11-4-2-3-7-6(5-9)8(10)13-12-7/h11H,2-4H2,1H3,(H3,10,12,13)
Inchi Key FJYUWIMEBHUCPT-UHFFFAOYSA-N

Description

Melting Point 127-128
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08532
Availability details for
Quantity
  • Availability
  • Add to Cart