ENA024294575 Enamine

7a-methyl-5-oxo-hexahydropyrrolo[2,1-b][1,3]thiazole-3-carboxylic acid

  • MDL number MFCD00553439
  • Cas Number : 17279-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08535
Availability details for
Quantity
  • Availability
  • Add to Cart