ENA024294581 Enamine

N-[2,6-bis(propan-2-yl)phenyl]-2-chloro-N-[4-(4-methylphenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD04620385
  • Cas Number : 727983-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08541
Availability details for
Quantity
  • Availability
  • Add to Cart