ENA024294585 Enamine

2-(1H-indol-3-yl)-2-(4-methoxyphenyl)ethan-1-amine

  • MDL number MFCD02731132
  • Cas Number : 203394-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08545
Availability details for
Quantity
  • Availability
  • Add to Cart