ENA024294589 Enamine

2-(2-chloro-1H-1,3-benzodiazol-1-yl)-1-phenylethan-1-one

  • MDL number MFCD00182104
  • Cas Number : 23085-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08549
Availability details for
Quantity
  • Availability
  • Add to Cart