ENA024294592 Enamine

1-[4-(pentyloxy)phenyl]ethanone

  • MDL number MFCD00027266
  • Cas Number : 5467-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08552

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08552
Availability details for
Quantity
  • Availability
  • Add to Cart