ENA024294604 Enamine

3-(1H-1,2,3,4-tetrazol-1-yl)benzene-1-sulfonamide

  • MDL number MFCD02703570
  • Cas Number : 326873-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08564
Availability details for
Quantity
  • Availability
  • Add to Cart