ENA024294607 Enamine

2-(4-cyanophenoxy)propanehydrazide

  • MDL number MFCD02254114
  • Cas Number : 438219-13-7
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-08568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08568
Availability details for
Quantity
  • Availability
  • Add to Cart