ENA024294619 Enamine

3-(3,5-dimethyl-1H-pyrazol-1-yl)propan-1-amine

  • MDL number MFCD00462177
  • Cas Number : 62821-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08580
Availability details for
Quantity
  • Availability
  • Add to Cart