ENA024294622 Enamine

3-[5-(dimethylsulfamoyl)-1-ethyl-1H-1,3-benzodiazol-2-yl]propanoic acid

  • MDL number MFCD04621665
  • Cas Number : 730951-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08583
Availability details for
Quantity
  • Availability
  • Add to Cart