ENA024294626 Enamine

2-{[(1-phenylethyl)amino]methyl}phenol

  • MDL number MFCD05263160
  • Cas Number : 128386-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08587

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08587
Availability details for
Quantity
  • Availability
  • Add to Cart