ENA024294631 Enamine

1-[(2,6-dichlorophenyl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD04621640
  • Cas Number : 730950-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08592

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08592
Availability details for
Quantity
  • Availability
  • Add to Cart