ENA024294641 Enamine

[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-amine

  • MDL number MFCD04621647
  • Cas Number : 3176-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08602
Availability details for
Quantity
  • Availability
  • Add to Cart