ENA024294649 Enamine

N-(8-chloro-2,3-dihydro[1,4]dioxino[2,3-g]quinoxalin-7-yl)benzenesulfonamide

  • MDL number MFCD02709822
  • Cas Number : 329269-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08610

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08610
Availability details for
Quantity
  • Availability
  • Add to Cart