ENA024294659 Enamine

1-(3,4-diethoxyphenyl)ethanone

  • MDL number MFCD00277131
  • Cas Number : 1137-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08620
Availability details for
Quantity
  • Availability
  • Add to Cart