ENA024294665 Enamine

2-(3,5-dichlorophenoxy)-5-(piperidin-1-ylsulfonyl)aniline

  • MDL number MFCD06655020
  • Cas Number : 851169-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08626

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08626
Availability details for
Quantity
  • Availability
  • Add to Cart